3-Acryloyl-2-oxazolidinone ≥98.0% (by GC)
Catalog # TCIAA1579-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Bromination Method):min. 96.0 %
- Melting point:81.0 to 85.0 deg-C
Specification Test Results
About this item
:
:141.13 g/mol
Storage Temperature:Refrigerator
CAS Number:2043-21-2
Formula:C₆H₇NO₃
MDL Number:MFCD04038407