4-Acetyl-4-phenylpiperidine hydrochloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA1570-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:239.74 g/mol
EINECS:233-694-0
CAS Number:10315-03-4
Formula:C₁₃H₁₇NO
MDL Number:MFCD00039037