Aminomethyl polystyrene resin 0.5 - 0.9 mmol/g, cross-linked with 1% DVB (200-400 mesh)
Catalog # TCIAA1543-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Loading:0.5 to 0.9 mmol/g
Specification Test Results
About this item
:
MDL Number:MFCD00130502