3-Amino-1H-pyrazole-4-carboxamide sulphate(2:1) ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:27511-79-1
:
Formula:C8H14N8O6S
MDL Number:MFCD00013093
:350.31 g/mol
EINECS:248-503-6
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Water:max. 5.0 %