Order Entry
Finland
ContactUsLinkComponent
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-glucopyranose ≥98.0% (by HPLC)
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-glucopyranose ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-glucopyranose ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC) (CAD):
    min. 98.0 area%
  • Specific rotation [a]20/D:
    +1.0 to +3.5 deg (C=5, CHCl3)
  • Solubility in Chloroform:
    almost transparency

Specification Test Results

Related Information