Order Entry
Finland
ContactUsLinkComponent
 
2-(2-Aminothiazol-4-yl)acetic acid ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
2-(2-Aminothiazol-4-yl)acetic acid ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %