
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one ≥98.0% (by titrimetric analysis)
Catalog # TCIAA0907-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Solubility in 1mol/L NaOH:almost transparency
Specification Test Results
About this item
:
:136.11 g/mol
Merck Index:14,00279
Melting Pt:384°C
EINECS:206-250-9
UN:2811
ADR:6.1,III
:GHS06
CAS Number:315-30-0
Formula:C₅H₄N₄O
MDL Number:MFCD00599413