4-(2-Acetoxyethoxy)toluene ≥98.0% (by GC)
Catalog # TCIAA0819-10G
Supplier: TCI
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White or Colorless to Light yellow to Light orange powder to lump to clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:194.23 g/mol
Boiling Pt:94°C
EINECS:229-881-1
CAS Number:6807-11-0
Formula:C₁₁H₁₄O₃
MDL Number:MFCD00059337