
Amyl methyl sulphide ≥97.0% (by GC)
Catalog # TCIAA0452-25ML
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
About this item
:
:118.24 g/mol
Boiling Pt:145°C
Melting Pt:-94°C
Melting Pt:–94°C
UN:1993
ADR:3,III
:GHS02
CAS Number:1741-83-9
Formula:C₆H₁₄S
MDL Number:MFCD00027274