5-Aminofluorescein (isomer I) ≥95.0% (by HPLC, titration analysis)
: TCI
:3326-34-9
:
:C₂₀H₁₃NO₅
:MFCD00005052
:347.33 g/mol
:222-043-6
:GHS07
- Appearance:Orange to Brown to Dark red powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 95.0 %