

Acenaphthylene ≥94.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:208-96-8
:
Formula:C₁₂H₈
MDL Number:MFCD00003806
:152.2 g/mol
Boiling Pt:275°C
Melting Pt:93°C
EINECS:205-917-1
:GHS07
:GHS08
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 94.0 %
- Melting point:87.0 to 94.0 deg-C