Order Entry
Finland
ContactUsLinkComponent
Calcium acetate xhydrate ≥93.5% (dry basis) analytical reagent
Calcium acetate xhydrate ≥93.5% (dry basis) analytical reagent
Catalog # 22268.264
Supplier:  VWR Chemicals
CAS Number:  
undefined
Calcium acetate xhydrate ≥93.5% (dry basis) analytical reagent
Catalog # 22268.264
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (calculated on dried substance):
    Min. 93.5 %
  • pH (20°C; 5 %):
    7.5 - 9.0
  • Heavy metals (as Pb):
    Max. 50 ppm
  • Cl (Chloride):
    Max. 100 ppm
  • SO4 (Sulphate):
    Max. 0.1 %
  • Fe (Iron):
    Max. 10 ppm
  • K (Potassium):
    Max. 0.1 %
  • Na (Sodium):
    Max. 0.1 %

Specification Test Results

About this item

 :
 :176.18 g/mol
Density:1.5 g/cm³ (20 °C)
EINECS:200-540-9
CAS Number:114460-21-8
 :5743-26-0
Formula:(H₃CCOO)₂Ca·nH₂O
MDL Number:MFCD00150019