Tellurium standard solution, 1000 mg/L Te in 5% nitric acid (from Te), standard for ICP, Specpure®
Molecular Formula: Te in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134062
: Traceable to NIST.
- MDL No.:MFCD00134062
Molecular Formula: Te in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134062