Order Entry
Finland
Orders LinkContactUsLinkComponent
G418 disulphate

G418 disulphate

Irritant
Supplier:   EUROCLONE
G418 disulphate
Supplier:  EUROCLONE
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate
  • CAS Number:108321-42-2
  • Formula:C₂₀H₄₄N₄O₁₈S₂
  • MDL Number:MFCD00058314
  • Molecular Weight:692.72 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each

  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.