Order Entry
Finland
ContactUsLinkComponent
G418 disulphate
G418 disulphate
Catalog # A2167.0001
Supplier:  PanReac AppliChem
CAS Number:  
G418 disulphate
Catalog # A2167.0001
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :692.72 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:108321-42-2
Formula:C₂₀H₄₄N₄O₁₈S₂
MDL Number:MFCD00058314