Order Entry
Finland
ContactUsLinkComponent
Xylenol orange tetrasodium salt, TECHNICAL
Xylenol orange tetrasodium salt, TECHNICAL
Catalog # 26086.103
Supplier:  VWR Chemicals
CAS Number:  
undefined
Xylenol orange tetrasodium salt, TECHNICAL
Catalog # 26086.103
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
 :760.59 g/mol
Melting Pt:∼210 °C
Storage Temperature:Ambient
EINECS:222-805-8
CAS Number:3618-43-7
 :313223-06-2
Formula:C₃₁H₂₈N₂Na₄O₁₃S
MDL Number:MFCD00044293