Devarda's alloy, granules, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:8049-11-4
:
MDL Number:MFCD00198176
Merck Index:13,02958
Boiling Pt:906 °C (1013 hPa)
Melting Pt:490…560 °C
Density:5.79 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3089
ADR:4.1,II
Specification Test Results
- N (Nitrogen):Max. 10 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
SDS_EN_CH_VWRC20942.pdf SDS_EN_DE_VWRC20942.pdf SDS_EN_NL_VWRC20942.pdf SDS_EN_UK_VWRC20942.pdf sds_VWRC20942
SDS
spec_VWRC20942.181 spec_VWRC20942.181 spec_VWRC20942.294