Order Entry
Finland
ContactUsLinkComponent
Surface active cleaning agent, Decon 75®
Surface active cleaning agent, Decon 75®
Catalog # 148-0322
Supplier:  DECON LAB
Surface active cleaning agent, Decon 75®
Catalog # 148-0322
Supplier:  DECON LAB
Specifications
  • Description:
    Surface active cleaning agent, Decon 75®
  • Pack type:
    Plastic bottle
  • Pk:
    5 L
Specifications
About this item
Surface active cleaning agent for laboratory, medical and specialised industrial applications.
Supplied as a liquid concentrate, for dilution with water. For `manual` cleaning, or for use in an ultrasonic cleaning bath.
PROPERTIES: Biodegradable, bactericidal, non-flammable and totally rinsable.
Delivery: This product is not available in all countries. Please check with your local Avantor International office or supplier.