Order Entry
Finland
ContactUsLinkComponent
5-Bromouridine-5′-triphosphate sodium salt (BrUTP ) 10 mM in TE buffer
Irritant
5-Bromouridine-5′-triphosphate sodium salt (BrUTP ) 10 mM in TE buffer
Catalog # 40026.
Supplier:  Biotium
CAS Number:  
Irritant
5-Bromouridine-5′-triphosphate sodium salt (BrUTP ) 10 mM in TE buffer
Catalog # 40026.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pack type:
    Plastic vial
  • Pk:
    25 µl
  • Seq:
    1166

Specifications

About this item

 :
 :628.98 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:161848-60-8
Formula:C₉H₁₁BrN₂Na₃O₁₅P₃