Order Entry
Finland
ContactUsLinkComponent
Human recombinant BDNF (from E. coli)
Human recombinant BDNF (from E. coli)
Certificates
undefined
Human recombinant BDNF (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity assay: Proliferation of a neuroblastoma cell line stably expressing TrkB (BR6), ED50 ≤40 ng/ml, Bioactivity ≥2,5×10⁴ units/mg
  • Form:
    Lyophilised
  • Gene ID:
    P23560
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    ANON2|brain-derived neurotrophic factor|BNDF|brain derived neurotrophic factor|BULN2
  • Protein/peptide name:
    BDNF
  • Purity:
    ≥95%
  • Molecular weight:
    13,6/27,3 kDa
  • Sequence:
    MHSDPARRGE LSVCDSISEW VTAADKKTAV DMSGGTVTVL EKVPVSKGQL KQYFYETKCN PMGYTKEGCR GIDKRHWNSQ CRTTQSYVRA LTMDSKKRIG WRFIRIDTSC VCTLTIKRGR
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Related Information