Order Entry
Finland
ContactUsLinkComponent
 
Human Recombinant IL-2 (Animal free) (from E. coli)
 
undefined
Human Recombinant IL-2 (Animal free) (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: CTLL-2 cell proliferation, ED50 ≤3 ng/ml, bioactivity ≥3,3×10⁵ units/mg
  • Gene ID:
    P60568
  • Reconstitution instructions:
    Sterile 10 mM HCl at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    TCGF|T cell growth factor|aldesleukin
  • UniProtKB:
    P60568
  • Protein/peptide name:
    IL-2
  • Purity:
    ≥97%
  • Molecular weight:
    15,5 kDa
  • Sequence:
    MAPTSSSTKK TQLQLEHLLL DLQMILNGIN NYKNPKLTRM LTFKFYMPKK ATELKHLQCL EEELKPLEEV LNLAQSKNFH LRPRDLISNI NVIVLELKGS ETTFMCEYAD ETATIVEFLN RWITFCQSII STLT
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay