Mouse Recombinant M-CSF (from E. coli)
About this item
Macrophage colony stimulating factor (M-CSF) is a hematopoietic growth factor that is widely produced by a variety of cells. M-CSF stimulates the proliferation and differentiation of hematopoietic stem cells into monocyte and macrophage cell types. M-CSF also acts through the colony stimulating factor 1 receptor (CSF1R) to modulate processes involved in immunology, bone metabolism, fertility, and pregnancy. Human M-CSF shows activity on mouse cells; however, mouse M-CSF shows no activity on human cells.
- High quality
- Low endotoxin
- Affordable
Caution: This product is for research use only and is not for use in humans.
4 Options Available Below
Product Details & Documents
Macrophage colony stimulating factor (M-CSF) is a hematopoietic growth factor that is widely produced by a variety of cells. M-CSF stimulates the proliferation and differentiation of hematopoietic stem cells into monocyte and macrophage cell types. M-CSF also acts through the colony stimulating factor 1 receptor (CSF1R) to modulate processes involved in immunology, bone metabolism, fertility, and pregnancy. Human M-CSF shows activity on mouse cells; however, mouse M-CSF shows no activity on human cells.
- High quality
- Low endotoxin
- Affordable
Caution: This product is for research use only and is not for use in humans.
Documents
Literature reference - PubMed ID 24960189
Additional Product Details
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SHBT200-08AF-10UG
- SHBT200-08AF-100UG
- SHBT200-08AF-500UG
- SHBT200-08AF-1MG
- Protein/peptide name:
- M-CSF (AF)
- M-CSF (AF)
- M-CSF (AF)
- M-CSF (AF)
- Protein synonyms:
- CSF-1|mgI-IM
- CSF-1|mgI-IM
- CSF-2|mgI-IM
- CSF-1|mgI-IM
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Mouse
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- P07141
- P07141
- P07141
- P07141
- Sequence:
- MKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKP
- MKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKP
- MKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKP
- MKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKP
- Protease-free:
- N
- N
- N
- N
- Animal-free:
- Y
- Y
- Y
- Y
- Nuclease-free:
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:
- Bioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mg
- Bioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mg
- Bioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mg
- Bioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mg
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5
- 10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5
- 10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5
- 10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5
- Molecular weight:
- 18,2/36,4 kDa
- 18,2/36,4 kDa
- 18,2/36,5 kDa
- 18,2/36,4 kDa
- Reconstitution instructions:
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Storage conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- Tested applications:
- Bioassay
- Bioassay
- Bioassay
- Bioassay
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SHBT200-08AF-10UGProtein/peptide nameM-CSF (AF)Protein synonymsCSF-1|mgI-IMProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP07141SequenceMKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKPProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mgPurity≥95%GradeRUOFormulation10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5Molecular weight18,2/36,4 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT200-08AF-100UGProtein/peptide nameM-CSF (AF)Protein synonymsCSF-1|mgI-IMProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP07141SequenceMKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKPProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mgPurity≥95%GradeRUOFormulation10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5Molecular weight18,2/36,4 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT200-08AF-500UGProtein/peptide nameM-CSF (AF)Protein synonymsCSF-2|mgI-IMProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP07141SequenceMKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKPProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mgPurity≥95%GradeRUOFormulation10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5Molecular weight18,2/36,5 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT200-08AF-1MGProtein/peptide nameM-CSF (AF)Protein synonymsCSF-1|mgI-IMProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP07141SequenceMKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKPProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mgPurity≥95%GradeRUOFormulation10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5Molecular weight18,2/36,4 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



