Human Recombinant G-CSF (Animal free) (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Persephin is a neurotrophic factor of the glial cell line-derived neurotrophic factor (GDNF) family. Persephin promotes survival and growth of dopaminergic and motor neurons, but not peripheral neurons. Persephin is a ligand for the RET receptor tyrosine kinase.
- High quality
- Low endotoxin
- Affordable
G-CSF synthesis occurs in monocyte, macrophage, epithelial, endothelial, and fibroblast cells after activation by bacterial endotoxins, tumor necrosis factor alpha (TNF-α).
The functional activity of G-CSF is mediated through the granulocyte colony-stimulating factor receptor (G-CSF-R) to activate JAK/STAT and MAPK signal transduction pathways. G-CSF also promotes neurogenesis and inhibits neuronal apoptosis. Human and mouse G-CSF proteins are cross-reactive.
Delivery: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: NFS-60 cell proliferation, ED50 ≤0,060 ng/ml, bioactivity ≥1,7×10⁷ units/mg
- Gene ID:P09919
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:GM-CSFβ|pluripoietin|MGI-1G|CSF-3
- UniProtKB:P09919
- Protein/peptide name:G-CSF
- Purity:≥95%
- Molecular weight:18,8 kDa
- Sequence:MTPLGPASSL PQSFLLKCLE QVRKIQGDGA ALQEKLCATY KLCHPEELVL LGHSLGIPWA PLSSCPSQAL QLAGCLSQLH SGLFLYQGLL QALEGISPEL GPTLDTLQLD VADFATTIWQ QMEELGMAPA LQPTQGAMPA FASAFQRRAG GVLVASHLQS FLEVSYRVLR HLAQP
- Endotoxin level:Low
- Formulation:20 mM acetic acid, 50 mM sodium chloride
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay