Order Entry
Finland
ContactUsLinkComponent
Acetone ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Acetone ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Catalog # HONC65749-195L
   Safer Human and Environmental Health
CAS Number:  
Acetone ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Catalog # HONC65749-195L
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Metal drum
  • Pk:
    195 L
  • Seq:
    5531
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.8 %
  • Boiling range:
    55.5 - 56.5 °C
  • Density (D 20/4):
    0.790 - 0.792
  • Refractive index (n 20/D):
    1.3580 - 1.3600
  • Non-volatile matter:
    Max. 10 ppm
  • Water insoluble substances:
    complying
  • Water (Karl Fischer):
    Max. 0.2 %
  • Free acid (as CH₃COOH):
    Max. 0.0015 %
  • Free alkali (as NH₃):
    Max. 0.0008 %
  • Acidity or alkalinity:
    complying
  • Aluminium (Al):
    Max. 0.00005 %
  • Boron (B):
    Max. 0.000002 %
  • Barium (Ba):
    Max. 0.00001 %
  • Bismuth (Bi):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.00005 %
  • Cadmium (Cd):
    Max. 0.000005 %
  • Cobalt (Co):
    Max. 0.000001 %
  • Chromium (Cr):
    Max. 0.000002 %
  • Copper (Cu):
    Max. 0.000002 %
  • Iron (Fe):
    Max. 0.00001 %
  • Potassium (K):
    Max. 0.00005 %
  • Lithium (Li):
    Max. 0.00001 %
  • Magnesium (Mg):
    Max. 0.00001 %
  • Manganese (Mn):
    Max. 0.000001 %
  • Molybdenum (Mo):
    Max. 0.00001 %
  • Sodium (Na):
    Max. 0.00005 %
  • Nickel (Ni):
    Max. 0.000002 %
  • Lead (Pb):
    Max. 0.000002 %
  • Tin (Sn):
    Max. 0.00001 %
  • Strontium (Sr):
    Max. 0.00001 %
  • Zinc (Zn):
    Max. 0.000005 %
  • KMnO₄ reducing matter (as O):
    Max. 0.0002 %
  • Reducing impurities:
    complying
  • Aldehydes (as HCHO):
    Max. 0.001 %
  • Benzene (GC):
    Max. 2 Vol.ppm
  • Ethanol (GC):
    Max. 0.01 %
  • Methanol (GC):
    Max. 0.05 Vol.%
  • 2-propanol (GC):
    Max. 0.05 Vol.%
  • Related substances (GC):
    complying
  • Appearance of the solution:
    complying
  • APHA:
    Max. 10
  • Solubility in water:
    complying

Specification Test Results

About this item

 :
 :58.08 g/mol
Boiling Pt:55…57 °C
Density:0.79 g/cm3
Flash Pt:–17 °C
UN:1090
ADR:3,II
 :GHS07
 :GHS02
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765