PTFE coated silicone seals, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
High-quality silicone seals developed for the needs of the pharmaceutical and biotech industry.
- Cleanroom packaged, supplied with Certificate of Analysis (CoA), USP <88> Class VI Pharmaceutical grade materials
- Recommended usable temperature range is –196 to 200 °C
- Sterilisable via steam/autoclave, chemical, or dry heat
- Lab washer compatible and microwave safe
High-quality, PTFE coated silicone seals are designed for use with DURAN® PURE Premium Screw Caps. The silicone portion is light ivory in color, while the PTFE portion is gray.
Packaging: 10 pieces supplied per bag.