Order Entry
Finland
ContactUsLinkComponent
Cut resistant gloves, Showa GP-KV1
Cut resistant gloves, Showa GP-KV1
Supplier:  Showa
Certificates
Cut resistant gloves, Showa GP-KV1
Supplier:  Showa
Specifications
  • Catalog No:
  • 112-6001
  • 112-6000
  • 112-5999
  • 112-6002
  • Model:
  • GP-KV1
  • GP-KV1
  • GP-KV1
  • GP-KV1
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Kevlar®
  • Kevlar®
  • Kevlar®
  • Kevlar®
  • Finish:
  • Latex coating on palm and fingertips
  • Latex coating on palm and fingertips
  • Latex coating on palm and fingertips
  • Latex coating on palm and fingertips
  • Gauge:
  • 10
  • 10
  • 10
  • 10
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (3X43C)
  • EN 388:2016 (3X43C)
  • EN 388:2016 (3X43C)
  • EN 388:2016 (3X43C)
Specifications