
Pentafluorobenzyl bromide ≥98.5% (by GC), LiChropur™ for GC derivatisation, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:1765-40-8
:
Formula:(C₆F₅)CH₂Br
MDL Number:MFCD00000299
:260.99 g/mol
Boiling Pt:174...175 °C (lit.)
Melting Pt:19...20 °C (lit.)
Density:1.728 g/m³ (25 °C) (lit.)
UN:3265
ADR:8,II
:GHS07
Specification Test Results
- Appearance (Color):Colorless to Faint Brown-Yellow
- Appearance (Form):Liquid or Solid
- Purity (GC Area %):≥ 98.5 %
- Refractive Index N20/D:1.470 - 1.472
- Proton NMR Spectrum:Conforms to Structure