Order Entry
Finland
ContactUsLinkComponent
Monensin methyl ester ≥97.0% (by TLC), Selectophore™ ionophore, Supelco®
Toxic
Monensin methyl ester ≥97.0% (by TLC), Selectophore™ ionophore, Supelco®
Catalog # 30552-100MG
Supplier:  Merck
CAS Number:  
undefined
Toxic
Monensin methyl ester ≥97.0% (by TLC), Selectophore™ ionophore, Supelco®
Catalog # 30552-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :684.908 g/mol
ADR:6.1,III
 :M-Clarity™ quality level = MQ100
UN:2811
 :GHS06
CAS Number:28636-21-7
Formula:C₃₇H₆₄O₁₁