
Aminoferrocene ≥98.0% (by titrimetric analysis) derivatisation reagent, for HPLC, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:1273-82-1
:
Formula:C₁₀H₂FeN
MDL Number:MFCD03844760
:191.97 g/mol
:GHS07
Storage Temperature:Freezer
Frequently Bought Together