Peqlab peqGOLD, DNA Isolation, Plant DNA Mini Kit
Supplier: VWR Chemicals
Certificates
About this item
Dependable isolation of high quality total cellular DNA from a variety of plant species and tissues. peqGOLD homogeniser columns remove cell debris and improve sample handling following lysis.
- Fast: Process 50 mg of fresh sample or 30 mg of dry sample in under 40 minutes
- Reproducible: Standardised method for reliable results from a variety of samples
- Safe: No organic extractions
- High quality: Purified DNA suitable for most downstream applications
Delivery: Each kit contains peqGOLD DNA mini columns, peqGOLD DNA homogeniser columns, 2 ml collection tubes, SP1 buffer, SP2 buffer, SP3 buffer, RNase A and elution buffer.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:Plant samples
- Target molecule:DNA
- Sample size:50 mg of fresh or 30 mg dry tissue
- Description:Plant DNA Mini Kit