Order Entry
Finland
ContactUsLinkComponent
Flexible, user-friendly liquid handling cobot for lab automation
Flexible, user-friendly liquid handling cobot for lab automation
Catalog # FLOW10-001
Supplier:  FLOW ROBOTICS
undefined
Flexible, user-friendly liquid handling cobot for lab automation
Catalog # FLOW10-001
Supplier:  FLOW ROBOTICS

Specifications

  • Description:
    Liquid handling cobot for laboratory automation
  • Seq:
    01
  • Pk:
    1

Specifications

About this item

A collaborative robot assisting you every day: flowbot ONE can be used by anyone in the lab - no programming experience is required. The technology behind flowbot ONE allows for the flexibility required in laboratories with shifting workflows e.g. R&D laboratories. Flexibility and adaptation to new lab procedures are cornerstones of the technological development of flowbot ONE. Whenever a procedure changes, the user can quickly set up flowbot ONE for use – enabling immediate automation adapted to the new procedures. The cobot has two pipette modules. A module can be a 1-channel, a 4-channel or an 8-channel pipette.

  • Easy to learn and requires no previous experience with pipetting robots
  • Common pipette tips with or without filter can be used
  • Controlled using an internet browser, no program installation is required
  • Program specification and execution logs can be saved and printed
  • Safety door detection and emergency stop button

Dimensions:
W×D×H (closed): 1000×600×800 mm
W×D×H (open): 1000×600×1100 mm

Weight: 70 kg
2 pipette modules: 1-channel, 4-channel and 8-channel

Use flowbot ONE in e.g.: R&D laboratories and analysis laboratories.

Use flowbot ONE for e.g.: Serial dilutions in plates, standard dilution series, preparation and dilution for analysis, e.g. HPLC, normalisation of samples – also when they come in different tubes, dispensing standards in test tubes, screening programs allowing control of addition of several reagents/ingredients.
For more specifications download the brochure (PDF).

Functions: General liquid transfer, serial dilutions, mixing, multiple dispensing, normalisation.

Certifications: Complies with the ISO 8655 standard.