Order Entry
Finland
ContactUsLinkComponent
Alcian blue solution used in nuclear staining, for histology, Sigma-Aldrich®
Alcian blue solution used in nuclear staining, for histology, Sigma-Aldrich®
Catalog # 1.01647.0500
Supplier:  Merck
CAS Number:  
undefined
Alcian blue solution used in nuclear staining, for histology, Sigma-Aldrich®
Catalog # 1.01647.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 ml
  • Seq:
    2725
  • Pk Info:
    Preparation Capacity 2000 - 3000 slides

Specifications

Specification Test Results

  • Suitability for microscopy:
    passes test
  • neutral mucopolysaccharides:
    purple
  • polysaccharides:
    purple
  • PAS negative acidic mucosubstances:
    light blue
  • Nuclei:
    Blue

Specification Test Results

About this item

 :
 :1298.86 g/mol
Density:1.006 g/cm3 (20 °C)
Storage Temperature:Ambient
 :1
CAS Number:33864-99-2
 :75881-23-1
Formula:C₅₆H₆₈Cl₄CuN₁₆S₄
MDL Number:MFCD00010720