Order Entry
Finland
ContactUsLinkComponent
Supelco® Organic Reference Standard, trans-2-Octenal
Supelco® Organic Reference Standard, trans-2-Octenal
Catalog # 52464-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, trans-2-Octenal
Catalog # 52464-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    2548-87-0
  • Description:
    trans-2-Octenal
  • Seq:
    01
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00007011
  • Formula:
    CH₃(CH₂)₄CH=CHCHO
  • Molecular weight:
    126,20
  • Density:
    0,846 g/ml at 25 °C (lit.)
  • Flash point:
    66 °C - Closed cup
  • Boiling point:
    84 - 86 °C/19 mmHg (lit.)

Specifications

About this item

  • GHS signal: GHS07
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338