Supelco® Organic Reference Standard, Protein Standard Mix 15 - 600 Kda
Supplier: Merck
Certificates
About this item
- GHS signal: GHS07
Caution: P261 - P264 - P273 - P280 - P301 + P312 - P302 + P352 + P312
Specifications
- Storage conditions:–20 °C
- Hazard class description:H302 + H312 + H332 - H334 - H412
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Flash point:Not applicable