Supelco® Organic Reference Standard, O-Succinyl-ʟ-Carnitine Lithium Salt
: Merck
- Formula:C₁₁H₁₉NO₆ ·xLi⁺
- Storage temperature:2...8 °C
- Description:O-Succinyl-L-carnitine lithium salt, ≥98,0% (TLC)
- Storage conditions:2...8 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Molecular weight:261,27 (free acid basis)
- Flash point:Not applicable
Frequently Bought Together