Order Entry
Finland
ContactUsLinkComponent
Organic reference standard, Monosodium N-lauroyl-L-glutamate, analytical standard, Supelco®
Organic reference standard, Monosodium N-lauroyl-L-glutamate, analytical standard, Supelco®
Catalog # 06927-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Monosodium N-lauroyl-L-glutamate, analytical standard, Supelco®
Catalog # 06927-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    29923-31-7
  • Description:
    Monosodium N-lauroyl-L-glutamate, ≥97,0% (TLC)
  • Formula:
    C₁₇H₃₀NNaO₅
  • Storage temperature:
  • Description:
    Monosodium N-lauroyl-L-glutamate

Specifications

About this item