Order Entry
Finland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Lipid Standards: Triglyceride Mixtures
Supelco® Organic Reference Standard, Lipid Standards: Triglyceride Mixtures
Catalog # 17811-1AMP
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Lipid Standards: Triglyceride Mixtures
Catalog # 17811-1AMP
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 Ampoul
  • Standards:
    Certified reference material
  • Description:
    Lipid standards: triglyceride mixtures
  • Storage conditions:
    –10...–25 °C
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ100
  • Storage temperature:
    –20 °C
  • Pk Info:
    100 mg

Specifications

About this item

Packaging: 100 mg ampule contains approximately equal amounts by weight of: Tricaprylin (C8:0) Tricaprin (C10:0) Trilaurin (C12:0) Trimyristin (C14:0) Tripalmitin (C16:0)