- Pk:100 mg
- Standards:Analytical standard
- CAS No.:105-60-2
- Description:epsilon-Caprolactam, ≥99,0% (GC)
- Environmentally Preferable:
- Melting point:68...71 °C (lit.)
- Formula:C₆H₁₁NO
- Description:epsilon-Caprolactam
- Flash point:152 °C
- Boiling point:136...138 °C/10 mmHg (lit.)
- Form:Neat
ε-Caprolactam is used in the manufacture of polyamides. It is also used to produce nylon 6.