Supelco® Organic Reference Standard, Cholesterol solution
Cholesterol solution, 10 mg/ml in chloroform, Certified reference material, 1
Catalog # 47127-U
Supplier: Merck
Supelco® Organic Reference Standard, Cholesterol solution
Catalog # 47127-U
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
- GHS signal: GHS06, GHS08
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Caution: P201 - P273 - P301 + P312 + P330 - P302 + P352 - P304 + P340 + P311 - P308 + P313
Specifications
Specifications
- Concentration:10 mg/ml in chloroform
- Pk:1
- Standards:Certified reference material
- Description:Cholesterol solution
- Cat. No.:47127-U
- M-Clarity quality level:MQ100
- Storage conditions:–10...–25 °C
- Hazard class description:H302 - H315 - H319 - H331 - H336 - H351 - H361d - H372 - H412
- MDL No.:MFCD00003646
- Molecular weight:386,65
- CAS No.:57-88-5
- Shipping conditions:STD
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



