Supelco® Organic Reference Standard, Asiatic acid
89773-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:464-92-6
- Description:Asiatic acid
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00238541
- M-Clarity quality level:MQ100
- Synonym(s):2α,23-Dihydroxyursolic acid; Dammarolic acid; NSC 166063
- Melting point:325 - 330 °C (lit.)
- Molecular weight:488,70