Order Entry
Finland
ContactUsLinkComponent
Organic reference standard, 3-Methyl-4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-3-buten-2-one, analytical standard, Supelco®
Organic reference standard, 3-Methyl-4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-3-buten-2-one, analytical standard, Supelco®
Catalog # 16797-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 3-Methyl-4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-3-buten-2-one, analytical standard, Supelco®
Catalog # 16797-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    127-51-5
  • Description:
    3-Methyl-4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-3-buten-2-one, ≥90,0% (GC)
  • Formula:
    C₁₄H₂₂O
  • Description:
    3-Methyl-4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-3-buten-2-one

Specifications

About this item