Order Entry
Finland
ContactUsLinkComponent
Organic reference standard, [D-Asp3, E-Dhb7]-Microcystin-RR solution, analytical standard, Supelco®
Organic reference standard, [D-Asp3, E-Dhb7]-Microcystin-RR solution, analytical standard, Supelco®
Catalog # 32541-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, [D-Asp3, E-Dhb7]-Microcystin-RR solution, analytical standard, Supelco®
Catalog # 32541-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 µg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    202120-08-9
  • Description:
    [D-Asp3, E-Dhb7]-Microcystin-RR solution
  • Formula:
    C₄₈H₇₃N₁₃O₁₂
  • Storage temperature:
    −20 °C
  • Description:
    [D-Asp3, E-Dhb7]-Microcystin-RR solution

Specifications

About this item

Microcystin is a type of toxin which can adversely affect human health. It is released by cyanobacteria and is commonly found in fresh water sources.

  • [D-Asp3, E-Dhb7]-Microcystin-RR may be used as an analytical standard for the determination of the analyte in:
  • Raw water and drinking water samples by ultra-high pressure liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
  • Water samples, Planktothrix cultures and European crayfish by LC-MS