Supelco® Organic Reference Standard, (RS)-(Methylenecyclopropyl)acetic acid
Catalog # 63978-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:1073-00-3
- Description:(RS)-(Methylenecyclopropyl)acetic acid
- Storage conditions:–20 °C
- Hazard class description:H302 - H315
- Shipping conditions:WICE
- MDL No.:MFCD11558999
- M-Clarity quality level:MQ100
- Molecular weight:112,13
Specifications
About this item
- GHS signal: GHS07