Lucifer Yellow Cadaverine Biotin-X dipotassium salt fluorescent dye, for histology
Catalog # 80017.
Supplier: Biotium
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
Storage Temperature:Refrigerator
Formula:C₃₃H₄₂K₂N₆O₁₁S₃