No. 2 Fuel oil, refinery grade, certified reference material, Supelco®
Catalog # 47515-U
Supplier: Merck
CAS Number:
Specifications
- Concentration:20000 mug/mL in methanol
- Pk:1
- Description:No. 2 Fuel Oil, refinery grade
- M-Clarity quality level:MQ100