Order Entry
Finland
ContactUsLinkComponent
Supelco® Organic Reference Standard, C8-C40 Alkanes Calibration Standard
Supelco® Organic Reference Standard, C8-C40 Alkanes Calibration Standard
Catalog # 40147-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, C8-C40 Alkanes Calibration Standard
Catalog # 40147-U
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Standards:
    In dichloromethane (varied)
  • Description:
    C8-C40 Alkanes Calibration Standard
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H315 - H319 - H336 - H351 - H373
  • Shipping conditions:
    STD
  • Concentration:
    In dichloromethane (varied)

Specifications

About this item

  • GHS signal: GHS08, GHS07
Caution: P202 - P260 - P264 - P302 + P352 - P305 + P351 + P338 - P308 + P313