About this item
Non woven wipes pre-saturated with acetone or isopropanol.
- Solvent-specific color-coded labeling for easy solvent identification
- Wipes are dispensed from center-pull perforated rolls with self-closing lids for added convenience
- Reusable canister is economical and solvent compatible
Contec's SATWipes® Solvent Wiping System consists of non woven pre-saturated wipes dispensed from a center-pull roll in a reusable canister. Custom engineered to meet a variety of needs, an assortment of fabrics, canisters and solvents are available. All fabrics and solvents are aerospace approved, and the canisters, pouches, and lids have been proven to withstand contact with the harshest of solvents. To preserve product integrity, SATWipes® rolls are packaged in specially designed foil pouches, which are opened and loaded into sturdy canisters. SATWipes® are tailored to specific tasks in any facility. This ensures that the right size wipe with the correct amount of solvent is used each time. Durable reusable canister with self-closing lid reduces waste, solvent use and VOCs.
Specifications
- Catalog No:
- CONTSW420195
- CONTSAT-C3-AC
- CONTSW420192
- Basis weight:
- 71 g/m²
- Extractables - using deionised water:
- 0,027 g/m²