Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Organo/Limonene Mount™
Organo/Limonene Mount™
Organo/Limonene Mount™
Organo/Limonene Mount™
Supplier:
SIGMA ALDRICH MICROSCOPY
Certificates
Expand image
Organo/Limonene Mount™
Supplier:
SIGMA ALDRICH MICROSCOPY
Certificates
About this item
:
Flash Pt:
48 °C
UN:
2052
ADR:
3,III
:
GHS02
:
GHS07
:
GHS08
:
GHS09
Related Information
Documentation
Certificates
Search by Lot Number
Product Literature
Merck The Color of Tissue Diagnostics
Documentation