




Chlorotrimethylsilane silylation reagent, for GC derivatisation
Supplier: MACHEREY-NAGEL
Certificates
About this item
CAS Number:75-77-4
:
Formula:C₃H₉ClSi
MDL Number:MFCD00000502
Boiling Pt:57 °C (760 mm Hg)
Density:0.8580 g/cm³ (d20°/4°)
:108.64 g/mol
:GHS02
:GHS05
:GHS06
:GHS07
:GHS08
Frequently Bought Together