O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride ≥99.0% (by argentometric titration), LiChropur™ for GC derivatisation, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:57981-02-9
:
Formula:C₇H₄F₅NO
MDL Number:MFCD00012953
:249.57 g/mol
:M-Clarity™ quality level = MQ100
Frequently Bought Together